missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL4R Protein
A cDNA sequence encoding the IL4R was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12425-3ug
Additional Details : Weight : 0.01000kg
Specifications
3566 | |
3 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
MKVLQEPTCV SDYMSISTCE WKMNGPTNCS TELRLLYQLV FLLSEAHTCI PENNGGAGCV CHLLMDDVVS ADNYTLDLWA GQQLLWKGSF KPSEHVKPRA PGNLTVHTNV SDTLLLTWSN PYPPDNYLYN HLTYAVNIWS ENDPADFRIY NVTYLEPSLR IAASTLKSGI SYRARVRAWAQCYNTTWSEW SPSTKWHNSY REPFEQHLEH HHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
IL4R Protein | |
Research Use Only | |
IL4R | |
Human | |
His | |
IL4R protein solution (0.25 mg/ml) contains Phosphate buffered saline (pH 7.4). |