missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL4R Protein
A cDNA sequence encoding the IL4R was constructed and used to recombinantly synthesize the protein.
£99.15 - £990.00
Specifications
Gene ID (Entrez) | 3566 |
---|---|
Name | IL4R Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | IL4R |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15980379
|
enQuireBio™
QP12425-3UG |
3 μg |
£99.15
3µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15970379
|
enQuireBio™
QP12425-15UG |
15 μg |
£155.00
15µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15960379
|
enQuireBio™
QP12425-0.1MG |
100 μg |
£990.00
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
3566 | |
Research Use Only | |
IL4R | |
Human | |
His | |
IL4R protein solution (0.25 mg/ml) contains Phosphate buffered saline (pH 7.4). |
IL4R Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
MKVLQEPTCV SDYMSISTCE WKMNGPTNCS TELRLLYQLV FLLSEAHTCI PENNGGAGCV CHLLMDDVVS ADNYTLDLWA GQQLLWKGSF KPSEHVKPRA PGNLTVHTNV SDTLLLTWSN PYPPDNYLYN HLTYAVNIWS ENDPADFRIY NVTYLEPSLR IAASTLKSGI SYRARVRAWAQCYNTTWSEW SPSTKWHNSY REPFEQHLEH HHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |