missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL4 / Interleukin-4 Protein

Recombinant Protein

Brand:  enQuireBio™ QP5233-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15935749

  • £3635.00 / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000580.1
IL4 / Interleukin-4 Protein
Research Use Only
IL4
Recombinant Protein
E. coli
MHKCDITLQE IIKTLNSLTE QKTLCTELTV TDIFAASKNT TEKETFCRAA TVLRQFYSHH EKDTRCLGAT AQQFHRHKQL IRFLKRLDRN LWGLAGLNSC PVKEANQSTL ENFLERLKTI MREKYSKCSS
Greater than 95% as determined by SDS-PAGE.
3565
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity at a concentration of 250 pg/mL in the TF-1 cell proliferation assay. The bioactivity acceptance criteria is set at 4 x 10^6 units/mg.
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers