missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL4 / Interleukin-4 Protein
Recombinant Protein
£2220.00 - £3635.00
Specifications
Accession Number | NP_000580.1 |
---|---|
Gene ID (Entrez) | 3565 |
Name | IL4 / Interleukin-4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15945749
|
enQuireBio™
QP5233-500UG |
500 μg |
£2220.00
500µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935749
|
enQuireBio™
QP5233-1MG |
1 mg |
£3635.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_000580.1 | |
IL4 / Interleukin-4 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 250 pg/mL in the TF-1 cell proliferation assay. The bioactivity acceptance criteria is set at 4 x 10^6 units/mg. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
3565 | |
Research Use Only | |
IL4 | |
Recombinant Protein | |
E. coli | |
MHKCDITLQE IIKTLNSLTE QKTLCTELTV TDIFAASKNT TEKETFCRAA TVLRQFYSHH EKDTRCLGAT AQQFHRHKQL IRFLKRLDRN LWGLAGLNSC PVKEANQSTL ENFLERLKTI MREKYSKCSS | |
Greater than 95% as determined by SDS-PAGE. |