missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL17RA Protein

A cDNA sequence encoding the IL17RA was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12408-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960329

  • £155.00 / 5µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

IL17RA Protein
Research Use Only
Recombinant Protein
Baculovirus
LRLLDHRALV CSQPGLNCTV KNSTCLDDSW IHPRNLTPSS PKDLQIQLHF AHTQQGDLFP VAHIEWTLQT DASILYLEGA ELSVLQLNTN ERLCVRFEFL SKLRHHHRRW RFTFSHFVVD PDQEYEVTVH HLPKPIPDGD PNHQSKNFLV PDCEHARMKV TTPCMSSGSL WDPNITVETL EAHQLRVSFT LWNESTHYQI LLTSFPHMEN HSCFEHMHHI PAPRPEEFHQ RSNVTLTLRN LKGCCRHQVQ IQPFFSSCLN DCLRHSATVS CPEMPDTPEP IPDYMPLWVE PKSCDKTHTC PPCPAPELLG GPSVFLFPPK PKDTLMISRT PEVTCVVVDV SHEDPEVKFN WYVDGVEVHN AKTKPREEQY NSTYRVVSVL TVLHQDWLNG KEYKCKVSNK ALPAPIEKTI SKAKGQPREP QVYTLPPSRD ELTKNQVSLT CLVKGFYPSD IAVEWESNGQ PENNYKTTPP VLDSDGSFFL YSKLTVDKSR WQQGNVFSCS VMHEALHNHY TQKSLSLSPG KHHHHHH
Greater than 90% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
IL17RA protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers