missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL17RA Protein
A cDNA sequence encoding the IL17RA was constructed and used to recombinantly synthesize the protein.
£99.15 - £1320.00
Specifications
Name | IL17RA Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15940329
|
enQuireBio™
QP12408-1UG |
1 μg |
£99.15
1µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15960329
|
enQuireBio™
QP12408-5UG |
5 μg |
£155.00
5µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950329
|
enQuireBio™
QP12408-50UG |
50 μg |
£1320.00
50µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
IL17RA Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
IL17RA protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
LRLLDHRALV CSQPGLNCTV KNSTCLDDSW IHPRNLTPSS PKDLQIQLHF AHTQQGDLFP VAHIEWTLQT DASILYLEGA ELSVLQLNTN ERLCVRFEFL SKLRHHHRRW RFTFSHFVVD PDQEYEVTVH HLPKPIPDGD PNHQSKNFLV PDCEHARMKV TTPCMSSGSL WDPNITVETL EAHQLRVSFT LWNESTHYQI LLTSFPHMEN HSCFEHMHHI PAPRPEEFHQ RSNVTLTLRN LKGCCRHQVQ IQPFFSSCLN DCLRHSATVS CPEMPDTPEP IPDYMPLWVE PKSCDKTHTC PPCPAPELLG GPSVFLFPPK PKDTLMISRT PEVTCVVVDV SHEDPEVKFN WYVDGVEVHN AKTKPREEQY NSTYRVVSVL TVLHQDWLNG KEYKCKVSNK ALPAPIEKTI SKAKGQPREP QVYTLPPSRD ELTKNQVSLT CLVKGFYPSD IAVEWESNGQ PENNYKTTPP VLDSDGSFFL YSKLTVDKSR WQQGNVFSCS VMHEALHNHY TQKSLSLSPG KHHHHHH | |
Greater than 90% as determined by SDS-PAGE. |