missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL17RA Protein
A cDNA sequence encoding the IL17RA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12408-1ug
Additional Details : Weight : 0.01000kg
Specifications
IL17RA Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
LRLLDHRALV CSQPGLNCTV KNSTCLDDSW IHPRNLTPSS PKDLQIQLHF AHTQQGDLFP VAHIEWTLQT DASILYLEGA ELSVLQLNTN ERLCVRFEFL SKLRHHHRRW RFTFSHFVVD PDQEYEVTVH HLPKPIPDGD PNHQSKNFLV PDCEHARMKV TTPCMSSGSL WDPNITVETL EAHQLRVSFT LWNESTHYQI LLTSFPHMEN HSCFEHMHHI PAPRPEEFHQ RSNVTLTLRN LKGCCRHQVQ IQPFFSSCLN DCLRHSATVS CPEMPDTPEP IPDYMPLWVE PKSCDKTHTC PPCPAPELLG GPSVFLFPPK PKDTLMISRT PEVTCVVVDV SHEDPEVKFN WYVDGVEVHN AKTKPREEQY NSTYRVVSVL TVLHQDWLNG KEYKCKVSNK ALPAPIEKTI SKAKGQPREP QVYTLPPSRD ELTKNQVSLT CLVKGFYPSD IAVEWESNGQ PENNYKTTPP VLDSDGSFFL YSKLTVDKSR WQQGNVFSCS VMHEALHNHY TQKSLSLSPG KHHHHHH | |
Greater than 90% as determined by SDS-PAGE. |
1 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
IL17RA protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |