missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL13RA1 Protein

A cDNA sequence encoding the IL13RA1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12405-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15920319

  • £155.00 / 10µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

3597
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
GGGGAAPTET QPPVTNLSVS VENLCTVIWT WNPPEGASSN CSLWYFSHFG DKQDKKIAPE TRRSIEVPLN ERICLQVGSQ CSTNESEKPS ILVEKCISPP EGDPESAVTE LQCIWHNLSY MKCSWLPGRN TSPDTNYTLY YWHRSLEKIH QCENIFREGQ YFGCSFDLTK VKDSSFEQHS VQIMVKDNAG KIKPSFNIVP LTSRVKPDPP HIKNLSFHND DLYVQWENPQ NFISRCLFYE VEVNNSQTET HNVFYVQEAK CENPEFERNV ENTSCFMVPG VLPDTLNTVR IRVKTNKLCY EDDKLWSNWS QEMSIGKKRN STHHHHHH
Greater than 90% as determined by SDS-PAGE.
IL13RA1 Protein
Research Use Only
IL13RA1
Human
His
IL13RA1 protein solution (0.5 mg/ml) in Phosphate buffer saline (pH 7.4).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers