missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL13RA1 Protein
A cDNA sequence encoding the IL13RA1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Gene ID (Entrez) | 3597 |
---|---|
Name | IL13RA1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | IL13RA1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15940319
|
enQuireBio™
QP12405-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15920319
|
enQuireBio™
QP12405-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15930319
|
enQuireBio™
QP12405-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
3597 | |
Research Use Only | |
IL13RA1 | |
Human | |
His | |
IL13RA1 protein solution (0.5 mg/ml) in Phosphate buffer saline (pH 7.4). |
IL13RA1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
GGGGAAPTET QPPVTNLSVS VENLCTVIWT WNPPEGASSN CSLWYFSHFG DKQDKKIAPE TRRSIEVPLN ERICLQVGSQ CSTNESEKPS ILVEKCISPP EGDPESAVTE LQCIWHNLSY MKCSWLPGRN TSPDTNYTLY YWHRSLEKIH QCENIFREGQ YFGCSFDLTK VKDSSFEQHS VQIMVKDNAG KIKPSFNIVP LTSRVKPDPP HIKNLSFHND DLYVQWENPQ NFISRCLFYE VEVNNSQTET HNVFYVQEAK CENPEFERNV ENTSCFMVPG VLPDTLNTVR IRVKTNKLCY EDDKLWSNWS QEMSIGKKRN STHHHHHH | |
Greater than 90% as determined by SDS-PAGE. |