missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL-35 Heterodimer Protein

Recombinant Protein

£77.85 - £2102.93

Specifications

Name IL-35 Heterodimer Protein
Product Type Recombinant Protein
Protein Tag Untagged
Host Species HEK 293
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15975659
View Documents Promotion Details
enQuireBio™
QP5203-2UG
2 μg
£77.85
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 18-02-2022
Please to purchase this item. Need a web account? Register with us today!
15965659
View Documents Promotion Details
enQuireBio™
QP5203-10UG
10 μg
£280.03
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 18-02-2022
Please to purchase this item. Need a web account? Register with us today!
15955659
View Documents Promotion Details
enQuireBio™
QP5203-100UG
100 μg
£2102.93
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 18-02-2022
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

IL-35 Heterodimer Protein
Untagged
Human
Greater than 95% as determined by SDS-PAGE.
Recombinant Protein
HEK 293
EBI3:RKGPP AALTLPRVQC RASRYPIAVD CSWTLPPAPN STSPVSFIAT YRLGMAARGH SWPCLQQTPT STSCTITDVQ LFSMAPYVLN VTAVHPWGSS SSFVPFITEH IIKPDPPEGV RLSPLAERQL QVQWEPPGSW PFPEIFSLKY WIRYKRQGAA RFHRVGPIEA TSFILRAVRP RARYYVQVAA QDLTDYGELS DWSLPATATM SLGK p35:RN LPVATPDPGM FPCLHHSQNL LRAVSNMLQK ARQTLEFYPC TSEEIDHEDI TKDKTSTVEA CLPLELTKNE SCLNSRETSF ITNGSCLASR KTSFMMALCL SSIYEDLKMY QVEFKTMNAK LLMDPKRQIF LDQNMLAVID ELMQALNFNS ETVPQKSSLE EPDFYKTKIK LCILLHAFRI RAVTIDRVMS YLNAS
IL35
SDS
Product Certifications