Learn More
enQuireBio™ Recombinant Human IL-35 Heterodimer Protein
Recombinant Protein
Brand:  enQuireBio™ QP5203-10ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15965659
  • £283.00 / 10µg
Estimated Shipment: 03-06-2024
to see stock.
Specifications
IL-35 Heterodimer Protein
Research Use Only
IL35
Human
Untagged
10 mM sodium phosphate, 30 mM sodium chloride, pH7.5
10 μg
Less than or equal to 5.00 EUs/ug as determined by the LAL method.
Recombinant Protein
Human (HEK293)
EBI3:RKGPP AALTLPRVQC RASRYPIAVD CSWTLPPAPN STSPVSFIAT YRLGMAARGH SWPCLQQTPT STSCTITDVQ LFSMAPYVLN VTAVHPWGSS SSFVPFITEH IIKPDPPEGV RLSPLAERQL QVQWEPPGSW PFPEIFSLKY WIRYKRQGAA RFHRVGPIEA TSFILRAVRP RARYYVQVAA QDLTDYGELS DWSLPATATM SLGK p35:RN LPVATPDPGM FPCLHHSQNL LRAVSNMLQK ARQTLEFYPC TSEEIDHEDI TKDKTSTVEA CLPLELTKNE SCLNSRETSF ITNGSCLASR KTSFMMALCL SSIYEDLKMY QVEFKTMNAK LLMDPKRQIF LDQNMLAVID ELMQALNFNS ETVPQKSSLE EPDFYKTKIK LCILLHAFRI RAVTIDRVMS YLNAS
Greater than 95% as determined by SDS-PAGE.