missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-29 / Interleukin-29 Protein
Recombinant Protein
Brand: enQuireBio™ QP5293-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_742152.1 | |
IL-29 / Interleukin-29 Protein | |
Research Use Only | |
IFNL1 | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
282618 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MPTSKPTTTG KGCHIGRFKS LSPQELASFK KARDALEESL KLKNWSCSSP VFPGNWDLRL LQVRERPVAL EAELALTLKV LEAAAGPALE DVLDQPLHTL HHILSQLQAC IQPQPTAGPR PRGRLHHWLH RLQEAPKKES AGCLEASVTF NLFRLLTRDL KYVADGNLCL RTSTHPEST | |
Greater than 95% as determined by SDS-PAGE. |