missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-29 / Interleukin-29 Protein
Recombinant Protein
£1890.00 - £3085.00
Specifications
Accession Number | NP_742152.1 |
---|---|
Gene ID (Entrez) | 282618 |
Name | IL-29 / Interleukin-29 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15975859
|
enQuireBio™
QP5293-500UG |
500 μg |
£1890.00
500µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965859
|
enQuireBio™
QP5293-1MG |
1 mg |
£3085.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_742152.1 | |
IL-29 / Interleukin-29 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MPTSKPTTTG KGCHIGRFKS LSPQELASFK KARDALEESL KLKNWSCSSP VFPGNWDLRL LQVRERPVAL EAELALTLKV LEAAAGPALE DVLDQPLHTL HHILSQLQAC IQPQPTAGPR PRGRLHHWLH RLQEAPKKES AGCLEASVTF NLFRLLTRDL KYVADGNLCL RTSTHPEST | |
Greater than 95% as determined by SDS-PAGE. |
282618 | |
Research Use Only | |
IFNL1 | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |