missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL-29 / Interleukin-29 Protein

Recombinant Protein

Brand:  enQuireBio™ QP5293-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15965859

  • £3085.00 / 1mg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_742152.1
IL-29 / Interleukin-29 Protein
Research Use Only
IFNL1
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
282618
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Recombinant Protein
E. coli
MPTSKPTTTG KGCHIGRFKS LSPQELASFK KARDALEESL KLKNWSCSSP VFPGNWDLRL LQVRERPVAL EAELALTLKV LEAAAGPALE DVLDQPLHTL HHILSQLQAC IQPQPTAGPR PRGRLHHWLH RLQEAPKKES AGCLEASVTF NLFRLLTRDL KYVADGNLCL RTSTHPEST
Greater than 95% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers