missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL 17A/F Protein
A cDNA sequence encoding the IL 17A/F was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10695-0.1mg
Additional Details : Weight : 0.01000kg
Spezifikation
IL 17A/F Protein | |
Research Use Only | |
The activity as determined by production of IL-6 from mouse 3T3 cells is 3.2ng/ml (3.1x105units/mg). | |
Human | |
Untagged | |
IL-17A/F was lyophilized from a (0.2microm) filtered protein solution containing 0.1% TFA. |
100 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MIVKAGITIP RNPGCPNSED KNFPRTVMVN LNIHNRNTNT NPKRSSDYYN RSTSPWNLHR NEDPERYPSV IWEAKCRHLG CINADGNVDY HMNSVPIQQE ILVLRREPPH CPNSFRLEKI LVSVGCTCVT PIVHHVAMRKIPKVGHT FFQKPESCPP VPGGSMKLDI GIINENQRVS MSRNIESRST SPWNYTVTWD PNRYPSEVVQ AQCRNLGCIN AQGKEDISMN SVPIQQETLV VRRKHQGCSV SFQLEKVLVT VGCTCVTPVI HHVQ | |
Greater than 95.0% as determined by SDS-PAGE. |