missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL 17A/F Protein
A cDNA sequence encoding the IL 17A/F was constructed and used to recombinantly synthesize the protein.
£99.15 - £990.00
Specifications
Name | IL 17A/F Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | The activity as determined by production of IL-6 from mouse 3T3 cells is 3.2ng/ml (3.1x105units/mg). |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15964678
|
enQuireBio™
QP10695-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954678
|
enQuireBio™
QP10695-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944678
|
enQuireBio™
QP10695-0.1MG |
100 μg |
£990.00
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
IL 17A/F Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MIVKAGITIP RNPGCPNSED KNFPRTVMVN LNIHNRNTNT NPKRSSDYYN RSTSPWNLHR NEDPERYPSV IWEAKCRHLG CINADGNVDY HMNSVPIQQE ILVLRREPPH CPNSFRLEKI LVSVGCTCVT PIVHHVAMRKIPKVGHT FFQKPESCPP VPGGSMKLDI GIINENQRVS MSRNIESRST SPWNYTVTWD PNRYPSEVVQ AQCRNLGCIN AQGKEDISMN SVPIQQETLV VRRKHQGCSV SFQLEKVLVT VGCTCVTPVI HHVQ | |
Greater than 95.0% as determined by SDS-PAGE. |
Research Use Only | |
The activity as determined by production of IL-6 from mouse 3T3 cells is 3.2ng/ml (3.1x105units/mg). | |
Human | |
Untagged | |
IL-17A/F was lyophilized from a (0.2microm) filtered protein solution containing 0.1% TFA. |