missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HSFY1 Protein
A cDNA sequence encoding the HSFY1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12324-20ug
Additional Details : Weight : 0.01000kg
Specifications
HSFY1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAHVSSE TQDVSPKDEL TASEASTRSP LCEHTFPGDS DLRSMIEEHA FQVLSQGSLL ESPSYTVCVS EPDKDDDFLS LNFPRKLWKI VESDQFKSIS WDENGTCIVI NEELFKKEIL ETKAPYRIFQ TDAIKSFVRQ LNLYGFSKIQ QNFQRSAFLA TFLSEEKESS VLSKLKFYYN PNFKRGYPQL LVRVKRRIGV KNASPISTLF NEDFNKKHFR AGANMENHNS ALAAEASEES LFSASKNLNM PLTRESSVRQ IIANSSVPIR SGFPPPSPST SVGPSEQIAT DQHAILNQLT TIHMHSHSTY MQARGHIVNF ITTTTSQYHI ISPLQNGYFG LTVEPSAVPT RYPLVSVNEA PYRNMLPAGN PWLQMPTIAD RSAAPHSRLA LQPSPLDKYH PNYN | |
Greater than 80.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The HSFY1 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |