missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HSFY1 Protein
A cDNA sequence encoding the HSFY1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | HSFY1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15980059
|
enQuireBio™
QP12324-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15970059
|
enQuireBio™
QP12324-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15960059
|
enQuireBio™
QP12324-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HSFY1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The HSFY1 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAHVSSE TQDVSPKDEL TASEASTRSP LCEHTFPGDS DLRSMIEEHA FQVLSQGSLL ESPSYTVCVS EPDKDDDFLS LNFPRKLWKI VESDQFKSIS WDENGTCIVI NEELFKKEIL ETKAPYRIFQ TDAIKSFVRQ LNLYGFSKIQ QNFQRSAFLA TFLSEEKESS VLSKLKFYYN PNFKRGYPQL LVRVKRRIGV KNASPISTLF NEDFNKKHFR AGANMENHNS ALAAEASEES LFSASKNLNM PLTRESSVRQ IIANSSVPIR SGFPPPSPST SVGPSEQIAT DQHAILNQLT TIHMHSHSTY MQARGHIVNF ITTTTSQYHI ISPLQNGYFG LTVEPSAVPT RYPLVSVNEA PYRNMLPAGN PWLQMPTIAD RSAAPHSRLA LQPSPLDKYH PNYN | |
Greater than 80.0% as determined by SDS-PAGE. |