missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HSFY1 Protein

A cDNA sequence encoding the HSFY1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12324-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960059

  • £2975.00 / 1mg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HSFY1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAHVSSE TQDVSPKDEL TASEASTRSP LCEHTFPGDS DLRSMIEEHA FQVLSQGSLL ESPSYTVCVS EPDKDDDFLS LNFPRKLWKI VESDQFKSIS WDENGTCIVI NEELFKKEIL ETKAPYRIFQ TDAIKSFVRQ LNLYGFSKIQ QNFQRSAFLA TFLSEEKESS VLSKLKFYYN PNFKRGYPQL LVRVKRRIGV KNASPISTLF NEDFNKKHFR AGANMENHNS ALAAEASEES LFSASKNLNM PLTRESSVRQ IIANSSVPIR SGFPPPSPST SVGPSEQIAT DQHAILNQLT TIHMHSHSTY MQARGHIVNF ITTTTSQYHI ISPLQNGYFG LTVEPSAVPT RYPLVSVNEA PYRNMLPAGN PWLQMPTIAD RSAAPHSRLA LQPSPLDKYH PNYN
Greater than 80.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The HSFY1 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers