missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HOXB13 Protein
A cDNA sequence encoding the HOXB13 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12296-20ug
Additional Details : Weight : 0.01000kg
Specifications
HOXB13 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEPGNYA TLDGAKDIEG LLGAGGGRNL VAHSPLTSHP AAPTLMPAVN YAPLDLPGSA EPPKQCHPCP GVPQGTSPAP VPYGYFGGGY YSCRVSRSSL KPCAQAATLA AYPAETPTAG EEYPSRPTEF AFYPGYPGTY QPMASYLDVS VVQTLGAPGE PRHDSLLPVD SYQSWALAGG WNSQMCCQGE QNPPGPFWKA AFADSSGQHP PDACAFRRGR KKRIPYSKGQ LRELEREYAA NKFITKDKRR KISAATSLSE RQITIWFQNR RVKEKKVLAK VKNSATP | |
Greater than 85.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
HOXB13 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |