missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HOXB13 Protein
A cDNA sequence encoding the HOXB13 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | HOXB13 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15989968
|
enQuireBio™
QP12296-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15979968
|
enQuireBio™
QP12296-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15969968
|
enQuireBio™
QP12296-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HOXB13 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
HOXB13 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEPGNYA TLDGAKDIEG LLGAGGGRNL VAHSPLTSHP AAPTLMPAVN YAPLDLPGSA EPPKQCHPCP GVPQGTSPAP VPYGYFGGGY YSCRVSRSSL KPCAQAATLA AYPAETPTAG EEYPSRPTEF AFYPGYPGTY QPMASYLDVS VVQTLGAPGE PRHDSLLPVD SYQSWALAGG WNSQMCCQGE QNPPGPFWKA AFADSSGQHP PDACAFRRGR KKRIPYSKGQ LRELEREYAA NKFITKDKRR KISAATSLSE RQITIWFQNR RVKEKKVLAK VKNSATP | |
Greater than 85.0% as determined by SDS-PAGE. |