missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Heterogeneous nuclear ribonucleoprotein D0 Protein
A DNA sequence encoding the Homo sapiens (Human) Heterogeneous nuclear ribonucleoprotein D0, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8416-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001003810.1 | |
Heterogeneous nuclear ribonucleoprotein D0 Protein | |
Research Use Only | |
HNRPD | |
Recombinant Protein | |
E. coli | |
AVGGSAGEQE GAMVAATQGA AAAAGSGAGT GGGTASGGTE GGSAESEGAK IDASKNEEDE GHSNSSPRHS EAATAQREEW KMFIGGLSWD TTKKDLKDYF SKFGEVVDCT LKLDPITGRS RGFGFVLFKE SESVDKVMDQ KEHKLNGKVI DPKRAKAMKT KEPVKKIFVG GLSPDTPEEK IREYFGGFGE VESIELPMDN KTNKRRGFCF ITFKEEEPVK KIMEKKYHNV GLSKCEIKVA MSKEQYQQQQ QWGSRGGFAG RARGRGGGPS QNWNQGYSNY WNQGYGNYGY NSQGYGGYGG YDYTGYNNYY GYGDYSNQQS GYGKVSRRGG HQNSYKPY | |
Greater than 90% as determined by SDS-PAGE. |
3184 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |