missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Heterogeneous nuclear ribonucleoprotein D0 Protein
A DNA sequence encoding the Homo sapiens (Human) Heterogeneous nuclear ribonucleoprotein D0, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001003810.1 |
---|---|
Gene ID (Entrez) | 3184 |
Name | Heterogeneous nuclear ribonucleoprotein D0 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16064491
|
enQuireBio™
QP8416-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16084491
|
enQuireBio™
QP8416-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16074491
|
enQuireBio™
QP8416-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16094491
|
enQuireBio™
QP8416-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16004501
|
enQuireBio™
QP8416-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16014501
|
enQuireBio™
QP8416-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001003810.1 | |
Heterogeneous nuclear ribonucleoprotein D0 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
3184 | |
Research Use Only | |
HNRPD | |
Recombinant Protein | |
E. coli | |
AVGGSAGEQE GAMVAATQGA AAAAGSGAGT GGGTASGGTE GGSAESEGAK IDASKNEEDE GHSNSSPRHS EAATAQREEW KMFIGGLSWD TTKKDLKDYF SKFGEVVDCT LKLDPITGRS RGFGFVLFKE SESVDKVMDQ KEHKLNGKVI DPKRAKAMKT KEPVKKIFVG GLSPDTPEEK IREYFGGFGE VESIELPMDN KTNKRRGFCF ITFKEEEPVK KIMEKKYHNV GLSKCEIKVA MSKEQYQQQQ QWGSRGGFAG RARGRGGGPS QNWNQGYSNY WNQGYGNYGY NSQGYGGYGG YDYTGYNNYY GYGDYSNQQS GYGKVSRRGG HQNSYKPY | |
Greater than 90% as determined by SDS-PAGE. |