missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HDL Protein

A cDNA sequence encoding the HDL was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12211-10mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15939708

  • £155.00 / 10mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HDL Protein
Research Use Only
Recombinant Protein
Human
Cholesterol level >200 mg/l. <1% LDL.
10 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers