missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HDL Protein
A cDNA sequence encoding the HDL was constructed and used to recombinantly synthesize the protein.
£155.00 - £5325.00
Specifications
Name | HDL Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15939708
|
enQuireBio™
QP12211-10MG |
10 mg |
£155.00
10mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15929708
|
enQuireBio™
QP12211-100MG |
100 mg |
£700.00
100mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949708
|
enQuireBio™
QP12211-1GRAM |
1 g |
£5325.00
1g |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HDL Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged |
Research Use Only | |
Recombinant Protein | |
Human | |
Cholesterol level >200 mg/l. <1% LDL. |