missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HDGF Protein
A cDNA sequence encoding the HDGF was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12205-5ug
Additional Details : Weight : 0.01000kg
Specifications
HDGF Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSRSNRQKEY KCGDLVFAKM KGYPHWPARI DEMPEAAVKS TANKYQVFFF GTHETAFLGP KDLFPYEESK EKFGKPNKRK GFSEGLWEIE NNPTVKASGY | |
Greater than 95% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The HDGF protein solution (1 mg/ml) is formulated in 20mM Tris-HCl pH 7.5, 1mM DTT, 10% glycerol and 1mM EDTA. |