missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HDGF Protein
A cDNA sequence encoding the HDGF was constructed and used to recombinantly synthesize the protein.
£99.15 - £3965.00
Specifications
Name | HDGF Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15969688
|
enQuireBio™
QP12205-5ug |
5 μg |
£99.15
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15959688
|
enQuireBio™
QP12205-20ug |
20 μg |
£155.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15949688
|
enQuireBio™
QP12205-1mg |
1 mg |
£3965.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
HDGF Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The HDGF protein solution (1 mg/ml) is formulated in 20mM Tris-HCl pH 7.5, 1mM DTT, 10% glycerol and 1mM EDTA. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSRSNRQKEY KCGDLVFAKM KGYPHWPARI DEMPEAAVKS TANKYQVFFF GTHETAFLGP KDLFPYEESK EKFGKPNKRK GFSEGLWEIE NNPTVKASGY | |
Greater than 95% as determined by SDS-PAGE. |