missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GNB3 Protein
A cDNA sequence encoding the GNB3 was constructed and used to recombinantly synthesize the protein.
£99.15 - £3965.00
Specifications
Name | GNB3 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15979098
|
enQuireBio™
QP12015-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15969098
|
enQuireBio™
QP12015-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15959098
|
enQuireBio™
QP12015-1MG |
1 mg |
£3965.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
GNB3 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
GNB3 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGEMEQL RQEAEQLKKQ IADARKACAD VTLAELVSGL EVVGRVQMRT RRTLRGHLAK IYAMHWATDS KLLVSASQDG KLIVWDSYTT NKVHAIPLRS SWVMTCAYAP SGNFVACGGL DNMCSIYNLK SREGNVKVSR ELSAHTGYLS CCRFLDDNNI VTSSGDTTCA LWDIETGQQK TVFVGHTGDC MSLAVSPDFN LFISGACDAS AKLWDVREGT CRQTFTGHES DINAICFFPN GEAICTGSDD ASCRLFDLRA DQELICFSHE SIICGITSVA FSLSGRLLFA GYDDFNCNVW DSMKSERVGI LSGHDNRVSC LGVTADGMAV ATGSWDSFLK IWN | |
Greater than 80.0% as determined by SDS-PAGE. |