missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GNB3 Protein
A cDNA sequence encoding the GNB3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12015-5ug
Additional Details : Weight : 0.01000kg
Specifications
GNB3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGEMEQL RQEAEQLKKQ IADARKACAD VTLAELVSGL EVVGRVQMRT RRTLRGHLAK IYAMHWATDS KLLVSASQDG KLIVWDSYTT NKVHAIPLRS SWVMTCAYAP SGNFVACGGL DNMCSIYNLK SREGNVKVSR ELSAHTGYLS CCRFLDDNNI VTSSGDTTCA LWDIETGQQK TVFVGHTGDC MSLAVSPDFN LFISGACDAS AKLWDVREGT CRQTFTGHES DINAICFFPN GEAICTGSDD ASCRLFDLRA DQELICFSHE SIICGITSVA FSLSGRLLFA GYDDFNCNVW DSMKSERVGI LSGHDNRVSC LGVTADGMAV ATGSWDSFLK IWN | |
Greater than 80.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
GNB3 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |