missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GDF-15 Protein
Recombinant Protein
Brand: enQuireBio™ QP5296-20ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004855.2 | |
GDF-15 Protein | |
Research Use Only | |
GDF15 | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
9518 | |
20 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MARNGDHCPL . GPGRCCRLH T. VRASLEDL GW. ADWVLSP REV. QVTMCI GACP. SQFRA ANMHA. QIKT SLHRLK. PDT VPAPCCV. PA SYNPMVLIQK TDTGVSLQ. T YDDLLAKDC. HCI | |
Greater than 95% as determined by SDS-PAGE. |