missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GDF-15 Protein
Recombinant Protein
£80.80 - £3965.00
Specifications
Accession Number | NP_004855.2 |
---|---|
Gene ID (Entrez) | 9518 |
Name | GDF-15 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15955869
|
enQuireBio™
QP5296-5UG |
5 μg |
£80.80
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935869
|
enQuireBio™
QP5296-20UG |
20 μg |
£191.00
20µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15915869
|
enQuireBio™
QP5296-100UG |
100 μg |
£525.00
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945869
|
enQuireBio™
QP5296-500UG |
500 μg |
£2425.00
500µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925869
|
enQuireBio™
QP5296-1MG |
1 mg |
£3965.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_004855.2 | |
GDF-15 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MARNGDHCPL . GPGRCCRLH T. VRASLEDL GW. ADWVLSP REV. QVTMCI GACP. SQFRA ANMHA. QIKT SLHRLK. PDT VPAPCCV. PA SYNPMVLIQK TDTGVSLQ. T YDDLLAKDC. HCI | |
Greater than 95% as determined by SDS-PAGE. |
9518 | |
Research Use Only | |
GDF15 | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |