missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GAGE12F Protein

A cDNA sequence encoding the GAGE12F was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11923-0.1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15958808

  • £1320.00 / 100µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

100008586; 645073; 26748; 2579
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSWRGRS TYYWPRPRRY VQPPEMIGPM RPEQFSDEVE PATPEEGEPA TQRQDPAAAQ EGEDEGASAG QGPKPEAHSQ EQGHPQTGCE CEDGPDGQEM DPPNPEEVKT PEEGEKQSQC
Greater than 85.0% as determined by SDS-PAGE.
GAGE12F Protein
Research Use Only
GAGE12F
Human
His
GAGE12F protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers