missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GAGE12F Protein
A cDNA sequence encoding the GAGE12F was constructed and used to recombinantly synthesize the protein.
£99.15 - £1320.00
Specifications
Gene ID (Entrez) | 100008586; 645073; 26748; 2579 |
---|---|
Name | GAGE12F Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | GAGE12F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15978808
|
enQuireBio™
QP11923-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15968808
|
enQuireBio™
QP11923-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15958808
|
enQuireBio™
QP11923-0.1MG |
100 μg |
£1320.00
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
100008586; 645073; 26748; 2579 | |
Research Use Only | |
GAGE12F | |
Human | |
His | |
GAGE12F protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 10% glycerol. |
GAGE12F Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSWRGRS TYYWPRPRRY VQPPEMIGPM RPEQFSDEVE PATPEEGEPA TQRQDPAAAQ EGEDEGASAG QGPKPEAHSQ EQGHPQTGCE CEDGPDGQEM DPPNPEEVKT PEEGEKQSQC | |
Greater than 85.0% as determined by SDS-PAGE. |