missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FOPNL Protein
A cDNA sequence encoding the FOPNL was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11897-5ug
Additional Details : Weight : 0.01000kg
Specifications
FOPNL Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MATVAELKAV LKDTLEKKGV LGHLKARIRA EVFNALDDDR EPRPSLSHEN LLINELIREY LEFNKYKYTA SVLIAESGQP VVPLDRQFLI HELNAFEESK DNTIPLLYGI LAHFLRGTKD GIQNAFLKGP SLQPSDPSLG RQPSRRKPMD DHLRKEEQKS TNIEDLHVSQ AVNR | |
Greater than 85% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The VAMP7 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 20% glycerol. |