missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FOPNL Protein
A cDNA sequence encoding the FOPNL was constructed and used to recombinantly synthesize the protein.
£99.15 - £3965.00
Specifications
Name | FOPNL Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15968728
|
enQuireBio™
QP11897-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15958728
|
enQuireBio™
QP11897-25UG |
25 μg |
£155.00
25µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15948728
|
enQuireBio™
QP11897-1MG |
1 mg |
£3965.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
FOPNL Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The VAMP7 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MATVAELKAV LKDTLEKKGV LGHLKARIRA EVFNALDDDR EPRPSLSHEN LLINELIREY LEFNKYKYTA SVLIAESGQP VVPLDRQFLI HELNAFEESK DNTIPLLYGI LAHFLRGTKD GIQNAFLKGP SLQPSDPSLG RQPSRRKPMD DHLRKEEQKS TNIEDLHVSQ AVNR | |
Greater than 85% as determined by SDS-PAGE. |