missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Fibronectin Protein

A cDNA sequence encoding the Fibronectin was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11881-10mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15938678

  • £1055.00 / 10mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Fibronectin Protein
Research Use Only
Recombinant Protein
Human
The Fibronectin was lyophilized from a concentrated 1 mg/ml solution containing 0.05M Tris Cl pH 7.5 and 0.1M NaCl.
10 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
Greater than 95.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers