missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fibronectin Protein
A cDNA sequence encoding the Fibronectin was constructed and used to recombinantly synthesize the protein.
£99.15 - £1055.00
Specifications
Name | Fibronectin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15958678
|
enQuireBio™
QP11881-200UG |
200 μg |
£99.15
200µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15948678
|
enQuireBio™
QP11881-1MG |
1 mg |
£160.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15938678
|
enQuireBio™
QP11881-10MG |
10 mg |
£1055.00
10mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Fibronectin Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Greater than 95.0% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Human | |
The Fibronectin was lyophilized from a concentrated 1 mg/ml solution containing 0.05M Tris Cl pH 7.5 and 0.1M NaCl. |