Learn More
enQuireBio™ Recombinant Human FGF8 Protein
Recombinant Protein
Brand:  enQuireBio™ QP5301-1mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15945879
  • £3085.00 / 1mg
Estimated Shipment: 12-06-2024
to see stock.
Specifications
NP_006110.1
FGF8 Protein
Research Use Only
FGF8
Recombinant Protein
E. coli
MQVTVQSSPN . FTQHVREQS L. VTDQLSRR LI. RTYQLYS RTS. GKHVQV LANK. RINAM AEDGD. PFAK LIVETD. TFG SRVRVRGAET GLYICMN. KK GKLIAKSN. G KGKDCVFTE.  IVLENNYTAL . QNAKYEGWY M. AFTRKGRP RK. GSKTRQH QRE. VHFMKR LPRG. HHTTE QSLRFEFLNY PPFTR. SLRG SQRTWA. PEP R
Greater than 95% as determined by SDS-PAGE.
2253
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
At minimum, our recombinant Human FGF8 induces 3T3 cell proliferation at concentrations of less than 150 ng/ml (activity must be at least 6.7 x 10^3 units/mg).
Human
Untagged
5 mM sodium phosphate, 50mM soldium chloride, pH 7.5