missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGF8 Protein
Recombinant Protein
£80.80 - £3085.00
Specifications
Accession Number | NP_006110.1 |
---|---|
Gene ID (Entrez) | 2253 |
Name | FGF8 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15975879
|
enQuireBio™
QP5301-5UG |
5 μg |
£80.80
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955879
|
enQuireBio™
QP5301-25UG |
25 μg |
£191.00
25µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935879
|
enQuireBio™
QP5301-100UG |
100 μg |
£476.00
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965879
|
enQuireBio™
QP5301-500UG |
500 μg |
£1890.00
500µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945879
|
enQuireBio™
QP5301-1MG |
1 mg |
£3085.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_006110.1 | |
FGF8 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
At minimum, our recombinant Human FGF8 induces 3T3 cell proliferation at concentrations of less than 150 ng/ml (activity must be at least 6.7 x 10^3 units/mg). | |
Human | |
Untagged | |
5 mM sodium phosphate, 50mM soldium chloride, pH 7.5 |
2253 | |
Research Use Only | |
FGF8 | |
Recombinant Protein | |
E. coli | |
MQVTVQSSPN . FTQHVREQS L. VTDQLSRR LI. RTYQLYS RTS. GKHVQV LANK. RINAM AEDGD. PFAK LIVETD. TFG SRVRVRGAET GLYICMN. KK GKLIAKSN. G KGKDCVFTE. IVLENNYTAL . QNAKYEGWY M. AFTRKGRP RK. GSKTRQH QRE. VHFMKR LPRG. HHTTE QSLRFEFLNY PPFTR. SLRG SQRTWA. PEP R | |
Greater than 95% as determined by SDS-PAGE. |