missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FAM84A Protein

A cDNA sequence encoding the FAM84A was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11849-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15928578

  • £155.00 / 20µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

FAM84A Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMGNQLDR ITHLNYSELP TGDPSGIEKD ELRVGVAYFF SDDEEDLDER GQPDKFGVKA PPGCTPCPES PSRHHHHLLH QLVLNETQFS AFRGQECIFS KVSGGPQGAD LSVYAVTALP ALCEPGDLLE LLWLQPAPEP PAPAPHWAVY VGGGQIIHLH QGEIRQDSLY EAGAANVGRV VNSWYRYRPL VAELVVQNAC GHLGLKSEEI CWTNSESFAA WCRFGKREFK AGGEVPAGTQ PPQQQYYLKV HLGENKVHTA RFHSLEDLIR EKRRIDASGR LRVLQELADL VDDKE
Greater than 90% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The FAM84A solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers