missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FAM84A Protein
A cDNA sequence encoding the FAM84A was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | FAM84A Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15938578
|
enQuireBio™
QP11849-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15928578
|
enQuireBio™
QP11849-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15918578
|
enQuireBio™
QP11849-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
FAM84A Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The FAM84A solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGNQLDR ITHLNYSELP TGDPSGIEKD ELRVGVAYFF SDDEEDLDER GQPDKFGVKA PPGCTPCPES PSRHHHHLLH QLVLNETQFS AFRGQECIFS KVSGGPQGAD LSVYAVTALP ALCEPGDLLE LLWLQPAPEP PAPAPHWAVY VGGGQIIHLH QGEIRQDSLY EAGAANVGRV VNSWYRYRPL VAELVVQNAC GHLGLKSEEI CWTNSESFAA WCRFGKREFK AGGEVPAGTQ PPQQQYYLKV HLGENKVHTA RFHSLEDLIR EKRRIDASGR LRVLQELADL VDDKE | |
Greater than 90% as determined by SDS-PAGE. |