missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FAM84A Protein
A cDNA sequence encoding the FAM84A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11849-1mg
Additional Details : Weight : 0.01000kg
Specifications
FAM84A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGNQLDR ITHLNYSELP TGDPSGIEKD ELRVGVAYFF SDDEEDLDER GQPDKFGVKA PPGCTPCPES PSRHHHHLLH QLVLNETQFS AFRGQECIFS KVSGGPQGAD LSVYAVTALP ALCEPGDLLE LLWLQPAPEP PAPAPHWAVY VGGGQIIHLH QGEIRQDSLY EAGAANVGRV VNSWYRYRPL VAELVVQNAC GHLGLKSEEI CWTNSESFAA WCRFGKREFK AGGEVPAGTQ PPQQQYYLKV HLGENKVHTA RFHSLEDLIR EKRRIDASGR LRVLQELADL VDDKE | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The FAM84A solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 10% glycerol. |