missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human F9 Protein

A cDNA sequence encoding the F9 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10595-100IU

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15944358

  • £990.00 / 100 units
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

F9 Protein
Research Use Only
The activity per mg was tested and found to be 306.5 PEU/mg.
Human
Untagged
Greater than 95.0% as determined by SDS-PAGE.
100 IU
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
Human
The Factor-IX was lyophilized from a sterile solution containing 20mM Tris-HCl pH 7.4, 0.1M NaCl and 1mM Benzamidine.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers