missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human F9 Protein
A cDNA sequence encoding the F9 was constructed and used to recombinantly synthesize the protein.
£990.00 - £7160.00
Specifications
Name | F9 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | The activity per mg was tested and found to be 306.5 PEU/mg. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15934358
|
enQuireBio™
QP10595-1000IU |
1000 IU |
£7160.00
1000 units |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944358
|
enQuireBio™
QP10595-100IU |
100 IU |
£990.00
100 units |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954358
|
enQuireBio™
QP10595-500IU |
500 IU |
£3965.00
500 units |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
F9 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Human | |
The Factor-IX was lyophilized from a sterile solution containing 20mM Tris-HCl pH 7.4, 0.1M NaCl and 1mM Benzamidine. |
Research Use Only | |
The activity per mg was tested and found to be 306.5 PEU/mg. | |
Human | |
Untagged | |
Greater than 95.0% as determined by SDS-PAGE. |