missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ErbB3 Protein

A cDNA sequence encoding the ErbB3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10592-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15944348

  • £3965.00 / 1mg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

ErbB3 Protein
Research Use Only
Measured by its ability to postpone tumor emerge time of spontaneous breast cancer in FVB/N transgenic mice and inhibit the development of tumor, effectively inhibit the growth of in situ transplanted breast cancer in FVB/N transgenic mice.
Human
Untagged
Each mg protein contains 1 mg aluminum hydroxide 10mM arginine, 10mM sodium chloride, 20mM sodium phosphate buffer and 5mM potassium phosphate.
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MRANDALQVL GLLFSLARGS EVGNSQAVCP GTLNGLSVTG DAENQYQTLY KLYERCEVVM GNLEIVLTGH NADLSFLQWI REVTGYVLVA MNEFSTLPLP NLRVVRGTQV YDGKFAIFVM LNYNTNSSHA LRQLRLTQLT EILSGGVYIE KNDKLCHMDT IDWRDIVRDR DAEIVVKDNG RSCPPCHEVC KGRCWGPGSE DCQTLTKTIC APQCNGHCFG PNPNQCCHDE CAGGCSGPQD TDCFACRHFN DSGACVPRCP QPLVYNKLTF QLEPNPHTKY QYGGVCVASC PHNFVVDQTS CVRACPPDKM EVDKNGLKMC EPCGGLCPKA CEGTGSGSRF QTVDSSNIDG FVNCTKILGN LDFLITGLNG DPWHKIPALD PEKLNVFRTV REITGYLNIQ SWPPHMHNFS VFSNLTTIGG RSLYNRGFSL LIMKNLNVTS LGFRSLKEIS AGRIYISANR QLCYHHSLNW TKVLRGPTEE RLDIKHNRPR RDCVAEGKVC DPLCSSGGCW GPGPGQCLSC RNYSRGGVCV THCNFLNGEP REFAHEAECF SCHPECQPME GTATCNGSGS DTCAQCAHFR DGPHCVSSCP HGVLGAKGPI YKYPDVQNEC RPCHENCTQG CKGPELQDCL GQTLVLIGKT HLTMALTVIA GLVVIFMMLG GTFLYWRGRR IQNKRAMRRY LERGESIEPL DPSEKANKVL ARIFKETELR KLKVLGSGVF GTVHKGVWIP EGESIKIPVC IKVIEDKSGR QSFQAVTDHM LAIGSLDHAH IVRLLGLCPG SSLQLVTQYL PLGSLLDHVR QHRGALGPQL LLNWGVQIAK GMYYLEEHGM VHRNLAARNV LLKSPSQVQV ADFGVADLLP PDDKQLLYSE AKTPIKWMAL ESIHFGKYTH QSDVWSYGVT VWELMTFGAE
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers