missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ErbB3 Protein
A cDNA sequence encoding the ErbB3 was constructed and used to recombinantly synthesize the protein.
£99.15 - £3965.00
Specifications
Name | ErbB3 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | Measured by its ability to postpone tumor emerge time of spontaneous breast cancer in FVB/N transgenic mice and inhibit the development of tumor, effectively inhibit the growth of in situ transplanted breast cancer in FVB/N transgenic mice. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15964348
|
enQuireBio™
QP10592-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954348
|
enQuireBio™
QP10592-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944348
|
enQuireBio™
QP10592-1MG |
1 mg |
£3965.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
ErbB3 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MRANDALQVL GLLFSLARGS EVGNSQAVCP GTLNGLSVTG DAENQYQTLY KLYERCEVVM GNLEIVLTGH NADLSFLQWI REVTGYVLVA MNEFSTLPLP NLRVVRGTQV YDGKFAIFVM LNYNTNSSHA LRQLRLTQLT EILSGGVYIE KNDKLCHMDT IDWRDIVRDR DAEIVVKDNG RSCPPCHEVC KGRCWGPGSE DCQTLTKTIC APQCNGHCFG PNPNQCCHDE CAGGCSGPQD TDCFACRHFN DSGACVPRCP QPLVYNKLTF QLEPNPHTKY QYGGVCVASC PHNFVVDQTS CVRACPPDKM EVDKNGLKMC EPCGGLCPKA CEGTGSGSRF QTVDSSNIDG FVNCTKILGN LDFLITGLNG DPWHKIPALD PEKLNVFRTV REITGYLNIQ SWPPHMHNFS VFSNLTTIGG RSLYNRGFSL LIMKNLNVTS LGFRSLKEIS AGRIYISANR QLCYHHSLNW TKVLRGPTEE RLDIKHNRPR RDCVAEGKVC DPLCSSGGCW GPGPGQCLSC RNYSRGGVCV THCNFLNGEP REFAHEAECF SCHPECQPME GTATCNGSGS DTCAQCAHFR DGPHCVSSCP HGVLGAKGPI YKYPDVQNEC RPCHENCTQG CKGPELQDCL GQTLVLIGKT HLTMALTVIA GLVVIFMMLG GTFLYWRGRR IQNKRAMRRY LERGESIEPL DPSEKANKVL ARIFKETELR KLKVLGSGVF GTVHKGVWIP EGESIKIPVC IKVIEDKSGR QSFQAVTDHM LAIGSLDHAH IVRLLGLCPG SSLQLVTQYL PLGSLLDHVR QHRGALGPQL LLNWGVQIAK GMYYLEEHGM VHRNLAARNV LLKSPSQVQV ADFGVADLLP PDDKQLLYSE AKTPIKWMAL ESIHFGKYTH QSDVWSYGVT VWELMTFGAE | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
Measured by its ability to postpone tumor emerge time of spontaneous breast cancer in FVB/N transgenic mice and inhibit the development of tumor, effectively inhibit the growth of in situ transplanted breast cancer in FVB/N transgenic mice. | |
Human | |
Untagged | |
Each mg protein contains 1 mg aluminum hydroxide 10mM arginine, 10mM sodium chloride, 20mM sodium phosphate buffer and 5mM potassium phosphate. |