missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ELAV-like protein 1
A DNA sequence encoding the Homo sapiens (Human) ELAV-like protein 4, was expressed in the hosts and tags indicated.
£145.46 - £1825.00
Specifications
Name | ELAV-like protein 1 |
---|---|
Product Type | Recombinant Protein |
Protein Tag | His-SUMO |
Host Species | E. coli |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15929119
|
enQuireBio™
QP5981-EC-100UG |
100 μg |
£400.00
100µg |
Estimated Shipment: 14-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15949119
|
enQuireBio™
QP5981-EC-1MG |
1 mg |
£1825.00
1mg |
Estimated Shipment: 14-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15939119
|
enQuireBio™
QP5981-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
15959119
|
enQuireBio™
QP5981-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
15969119
|
enQuireBio™
QP5981-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
15979119
|
enQuireBio™
QP5981-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
Specifications
ELAV-like protein 1 | |
His-SUMO | |
Human | |
Greater than 80% as determined by SDS-PAGE. | |
NP_001138246.1 |
Recombinant Protein | |
E. coli | |
MVMIISTMEP QVSNGPTSNT SNGPSSNNRN CPSPMQTGAT TDDSKTNLIV NYLPQNMTQE EFRSLFGSIG EIESCKLVRD KITGQSLGYG FVNYIDPKDA EKAINTLNGL RLQTKTIKVS YARPSSASIR DANLYVSGLP KTMTQKELEQ LFSQYGRIIT SRILVDQVTG VSRGVGFIRF DKRIEAEEAI KGLNGQKPSG ATEPITVKFA NNPSQKSSQA LLSQLYQSPN RRYPGPLHHQ AQRFRLDNLL NMAYGVKRLM SGPVPPSACP PRFSPITIDG MTSLVGMNIP GHTGTGWCIF VYNLSPDSDE SVLWQLFGPF GAVNNVKVIR DFNTNKCKGF GFVTMTNYDE AAMAIASLNG YRLGDRVLQV SFKTNKAHKS | |
1996 | |
ELAVL4 |