missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ELAV-like protein 1
A DNA sequence encoding the Homo sapiens (Human) ELAV-like protein 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5981-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001138246.1 | |
ELAV-like protein 1 | |
Research Use Only | |
ELAVL4 | |
Recombinant Protein | |
E. coli | |
MVMIISTMEP QVSNGPTSNT SNGPSSNNRN CPSPMQTGAT TDDSKTNLIV NYLPQNMTQE EFRSLFGSIG EIESCKLVRD KITGQSLGYG FVNYIDPKDA EKAINTLNGL RLQTKTIKVS YARPSSASIR DANLYVSGLP KTMTQKELEQ LFSQYGRIIT SRILVDQVTG VSRGVGFIRF DKRIEAEEAI KGLNGQKPSG ATEPITVKFA NNPSQKSSQA LLSQLYQSPN RRYPGPLHHQ AQRFRLDNLL NMAYGVKRLM SGPVPPSACP PRFSPITIDG MTSLVGMNIP GHTGTGWCIF VYNLSPDSDE SVLWQLFGPF GAVNNVKVIR DFNTNKCKGF GFVTMTNYDE AAMAIASLNG YRLGDRVLQV SFKTNKAHKS | |
Greater than 80% as determined by SDS-PAGE. |
1996 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |