missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EGF Protein
A cDNA sequence encoding the EGF was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10582-100ug
Additional Details : Weight : 0.01000kg
Specifications
EGF Protein | |
Research Use Only | |
Recombinant Protein | |
P. pastoris | |
NSDSECPLSH DGYCLHDGVC MYIEALDKYA CNCVVGYIGE RCQYRDLKWW ELR | |
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
100 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Human | |
Untagged | |
Each lyophilized mg contains 0.15M NaCl, 0.025M sodium bicarbonate, pH 7.5. |