missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EGF Protein
A cDNA sequence encoding the EGF was constructed and used to recombinantly synthesize the protein.
£99.15 - £283.00
Specifications
Name | EGF Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15944318
|
enQuireBio™
QP10582-100UG |
100 μg |
£99.15
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964318
|
enQuireBio™
QP10582-500UG |
500 μg |
£155.00
500µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954318
|
enQuireBio™
QP10582-1MG |
1 mg |
£283.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
EGF Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Human | |
Untagged | |
Each lyophilized mg contains 0.15M NaCl, 0.025M sodium bicarbonate, pH 7.5. |
Research Use Only | |
Recombinant Protein | |
P. pastoris | |
NSDSECPLSH DGYCLHDGVC MYIEALDKYA CNCVVGYIGE RCQYRDLKWW ELR | |
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |